Learn More
N-Biotinyl-12-aminododecanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | N-Biotinyl-12-aminododecanoic Acid |
| CAS | 135447-73-3 |
| Molecular Formula | C22 H39 N3 O4 S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C22H39N3O4S/c26-19(13-10-9-12-18-21-17(16-30-18)24-22(29)25-21)23-15-11-7-5-3-1-2-4-6-8-14-20(27)28/h17-18,21H,1-16H2,(H,23,26)(H,27,28)(H2,24,25,29)/t17-,18-,21-/m0/s1 |
| SMILES | OC(=O)CCCCCCCCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12 |
| IUPAC Name | 12-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]dodecanoic acid |
| Molecular Weight (g/mol) | 441.63 |
| Formula Weight | 441.2661 |