Learn More
N-Allylthiourea, 98%
Product Code. p-7043899
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11332126
Brand: Thermo Scientific Alfa Aesar L03377.14
Request Bulk or Custom Format
CAS: 109-57-9 | C4H8N2S | 116.182 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
N-Allylthiourea is a nitrification inhibitor used in the study on the transformation of diclofenac, naproxen and bisoprolol under aerobic and anaerobic conditions. It is also used in medicine to minimize scar tissue in order to fight against a type of dermatitis. Further, it inhibits the growth of transplanted tumors in mice. It acts as a chelating agent. In addition, it is used in cosmetics, preservative and in organic synthesis.
Solubility
Soluble in water and alcohol. Slightly soluble in diethyl ether. Insoluble in benzene.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109-57-9
Molecular Formula C4H8N2S
Molecular Weight (g/mol) 116.182
MDL Number MFCD00004940
InChI Key HTKFORQRBXIQHD-UHFFFAOYSA-N
Synonym allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
PubChem CID 1549517
ChEBI CHEBI:74079
IUPAC Name prop-2-enylthiourea
SMILES C=CCNC(=S)N
Specifications
Melting Point 70°C to 78°C
Density 1.22
Odor Unpleasant
Quantity 25 g
UN Number UN2811
Beilstein 1071470
Merck Index 14,9362
Solubility Information Soluble in water and alcohol. Slightly soluble in diethyl ether. Insoluble in benzene.
Formula Weight 116.19
Percent Purity 98%
Chemical Name or Material N-Allylthiourea