Learn More
N-Acetyl-D-serine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-D-serine |
| CAS | 152612-69-6 |
| Molecular Formula | C5H9NO4 |
| Quantity | 500 mg |
| Synonym | (R)-(+)-2-(N-Acetylamino)-3-hydroxypropanoic Acid |
| InChI Formula | InChI=1S/C5H9NO4/c1-3(8)6-4(2-7)5(9)10/h4,7H,2H2,1H3,(H,6,8)(H,9,10)/t4-/m1/s1 |
| SMILES | CC(=O)N[C@H](CO)C(=O)O |
| IUPAC Name | (2R)-2-acetamido-3-hydroxypropanoic acid |
| Molecular Weight (g/mol) | 147.13 |
| Formula Weight | 147.0532 |