missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-D-glucosamine, 100%, MP Biomedicals™

Product Code. 11308359 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11308359

Brand: MP Biomedicals 0210006825

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00061615
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
Synonym 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Color White
Assay Percent Range 1
Quantity 25 g
Formula Weight 221.2
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Acetyl-D-glucosamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.