Learn More
N-Acetyl-3-acetoxyindole, TRC
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-3-acetoxyindole |
| CAS | 16800-67-2 |
| Molecular Formula | C12 H11 N O3 |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C12H11NO3/c1-8(14)13-7-12(16-9(2)15)10-5-3-4-6-11(10)13/h3-7H,1-2H3 |
| SMILES | CC(=O)Oc1cn(C(=O)C)c2ccccc12 |
| IUPAC Name | (1-acetylindol-3-yl) acetate |
| Molecular Weight (g/mol) | 217.22 |
| Formula Weight | 217.0739 |