Learn More
N-(4-Bromophenyl)maleimide, 98%
Product Code. 11458780
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458780 1 g 1g
11474464 5 g 5g
11484464 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11458780 Supplier Thermo Scientific Alfa Aesar Supplier No. A13119.03
Request Bulk or Custom Format
CAS: 13380-67-1 | C10H6BrNO2 | 252.067 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13380-67-1
Molecular Formula C10H6BrNO2
Molecular Weight (g/mol) 252.067
MDL Number MFCD00030664
InChI Key FECSFBYOMHWJQG-UHFFFAOYSA-N
Synonym n-4-bromophenyl maleimide, 1-4-bromophenyl-1h-pyrrole-2,5-dione, 1-4-bromophenyl pyrrole-2,5-dione, 1-4-bromophenyl-2,5-dioxo-pyrrole, 1-4-bromophenyl maleimide, 1-4-bromophenyl-2,5-dihydro-1h-pyrrole-2,5-dione, 1-4-bromphenyl-1h-pyrrole-2,5-dione, 1h-pyrrole-2,5-dione,1-4-bromophenyl, 1h-pyrrole-2,5-dione, 1-4-bromophenyl, 1-4-bromophenyl azoline-2,5-dione
PubChem CID 123301
IUPAC Name 1-(4-bromophenyl)pyrrole-2,5-dione
SMILES C1=CC(=CC=C1N2C(=O)C=CC2=O)Br
Specifications
Melting Point 127°C to 131°C
Quantity 1 g
Beilstein 513122
Formula Weight 252.07
Percent Purity 98%
Chemical Name or Material N-(4-Bromophenyl)maleimide