missing translation for 'onlineSavingsMsg'
Learn More

N-(3,4-Dimethoxyphenethyl)formamide, TRC

Product Code. 30378107
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378107 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378107 Supplier Toronto Research Chemicals Supplier No. TRCD4724602.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-(3,4-Dimethoxyphenyl)ethyl]formamide
CAS 14301-36-1
Molecular Formula C11 H15 N O3
Quantity 2.5 g
Synonym Formamide, N-[2-(3,4-dimethoxyphenyl)ethyl]-, Formamide, N-(3,4-dimethoxyphenethyl)- (6CI,7CI), N-[2-(3,4-Dimethoxyphenyl)ethyl]formamide, N-Formyl-2-(3,4-dimethoxyphenyl)ethylamine, NSC 34982, N-(3,4-Dimethoxyphenethyl)formamide
InChI Formula InChI=1S/C11H15NO3/c1-14-10-4-3-9(5-6-12-8-13)7-11(10)15-2/h3-4,7-8H,5-6H2,1-2H3,(H,12,13)
SMILES COc1ccc(CCNC=O)cc1OC
IUPAC Name N-[2-(3,4-dimethoxyphenyl)ethyl]formamide
Molecular Weight (g/mol) 209.242
Formula Weight 209.105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.