Learn More
Macherey Nagel X100 N 20 Magnetic crimp cap, silver, 8 mm center hole
Brand:  Macherey Nagel 702808
X100 N 20 Magnetic crimp cap, silver, 8 mm center hole
Product Code. 12794162
  • ONLINE EXCLUSIVE PRICE
    9.55 EUR
    9.20 EUR / Pack of 100
    Save 0.35 EUR
    3% Off
Only null left
Estimated Shipment: 21-01-2026