Learn More
Macherey-Nagel™ N 20 Bi-metal Crimp Caps
Shop All Macherey-Nagel Products
Click to view available options
Septum:
Butyl/PTFE
Silicone/PTFE
Color (Seal):
Light Gray/Dark Gray
Transparent Blue/Clear
White/Beige
Unit Size:
Pack of 100
Specifications
Specifications
| Color (Seal) | Light Gray/Dark Gray |
| Color | Blue, Silver |
| Material | Bi-metal, Butyl Rubber, PTFE |
| Septum | Butyl/PTFE |
| Thickness (Metric) | 3 mm |