missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Aminoethyl)maleimide Trifluoroacetic Acid Salt, TRC

Product Code. 30010822
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010822 500 mg 500mg
30005373 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010822 Supplier Toronto Research Chemicals Supplier No. TRCA609050500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146474-00-2
Molecular Formula C6 H8 N2 O2 . C2 H F3 O2
Molecular Weight (g/mol) 254.16
Synonym 1H-Pyrrole-2,5-dione, 1-(2-aminoethyl)-, 2,2,2-trifluoroacetate (1:1), 1H-Pyrrole-2,5-dione, 1-(2-aminoethyl)-, mono(trifluoroacetate) (9CI), 1-(2-Aminoethyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate, 1-(2-Aminoethyl)-1H-pyrrole-2,5-dione trifluoroacetate, 1-(2-Aminoethyl)-2,5-dihydro-1H-pyrrole-2,5-dione, trifluoroacetic acid, 2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethananinium 2,2,2-trifluoroacetate, 2-Maleimidoethylamine trifluoroacetate, N-(2-Aminoethyl)maleimide trifluoracetate salt, N-(2-Aminoethyl)maleimide trifluoroacetate, N-(2-Aminoethyl)maleimide trifluoroacetate salt
IUPAC Name 1-(2-aminoethyl)pyrrole-2,5-dione;2,2,2-trifluoroacetic acid
SMILES NCCN1C(=O)C=CC1=O.OC(=O)C(F)(F)F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H8N2O2.C2HF3O2/c7-3-4-8-5(9)1-2-6(8)10;3-2(4,5)1(6)7/h1-2H,3-4,7H2;(H,6,7)
Formula Weight 254.0514
Percent Purity >95%
Chemical Name or Material N-(2-Aminoethyl)maleimide Trifluoroacetic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.