Learn More
Macherey-Nagel™ N 11 Aluminum Crimp Caps
Shop All Macherey-Nagel Products
Click to view available options
Color:
Blue
Red
Silver
Septum:
Natural Rubber/Butyl/TEF
Natural rubber / Butyl red orange/ TEF colorless
PTFE/Silicone/PTFE
Silicone/PTFE
Silicone/PTFE, Pre-slit
Virgin PTFE
Color (Seal):
Red/Orange/Clear
Red/White/Red
White
White/Blue
White/Red
Thickness (Metric):
0.25 mm
1 mm
1.3 mm
1.5 mm
Pakningsstørrelse:
Pack of 100
Beskrivelse
- Ready assembled crimp closures and plain crimp caps N 11
- Septa are assembled, caps are ready-to-use.
- Septa are shown for illustration purposes only, images upside down.
- Packs of 100 closures each
Tekniske data
Tekniske data
| Color (Seal) | White |
| Color | Silver |
| Material | Aluminum, PTFE |
| Septum | Virgin PTFE |
| Quantity | 100 Pk. |
| Thickness (Metric) | 0.25 mm |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion