missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYPN Antibody (4C8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00084665-M04
This item is not returnable.
View return policy
Description
MYPN Monoclonal antibody specifically detects MYPN in Human samples. It is validated for Western Blot, ELISA
Specifications
| MYPN | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| 145 kDa (MYOP), MYOP145 kDa sarcomeric protein, myopalladin | |
| MYPN (NP_115967, 61 a.a. ∽ 170 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PDLSAFLSQEELDESVNLARLAINYDPLEKADETQARKRLSPDQMKHSPNLSFEPNFCQDNPRSPTSSKESPQEAKRPQYCSETQSKKVFLNKAADFIEELSSLFKSHSS | |
| 0.1 mg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 84665 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 4C8 | |
| Western Blot 1:500, ELISA | |
| NP_115967 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction