missing translation for 'onlineSavingsMsg'
Learn More

MYOT Monoclonal Antibody (OTI3D1), TrueMAB™, OriGene

Product Code. 17561964
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17561964 30 μL 30µL
17541964 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17561964 Supplier Origene Technologies Supplier No. TA809573S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MYOT Monoclonal antibody specifically detects MYOT in Human samples. It is validated for Western Blot

Specifications

Antigen MYOT
Applications Western Blot
Classification Monoclonal
Clone OTI3D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RORC
Gene Accession No. P51449
Gene Alias 5530402I04Rik; 57 kDa cytoskeletal protein; LGMD1; LGMD1A; MFM3; Myo; Myofibrillar titin-like Ig domains protein; MYOT; Myotilin; titan-like protein; titin immunoglobulin domain protein; titin immunoglobulin domain protein (myotilin); Ttid; TTOD
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-266 of human RORC produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6097
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.