missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYO18A Polyclonal Antibody
GREENER_CHOICE

Product Code. 16622805 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16622805 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16622805 Supplier Invitrogen™ Supplier No. PA5106364

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MYO18A.

MYO18A may link Golgi membranes to the cytoskeleton and participate in the tensile force required for vesicle budding from the Golgi. Thereby, may play a role in Golgi membrane trafficking and could indirectly give its flattened shape to the Golgi apparatus. Alternatively, in concert with LURAP1 and CDC42BPA/CDC42BPB, has been involved in modulating lamellar actomyosin retrograde flow that is crucial to cell protrusion and migration. May be involved in the maintenance of the stromal cell architectures required for cell to cell contact. Regulates trafficking, expression, and activation of innate immune receptors on macrophages. Plays a role to suppress inflammatory responsiveness of macrophages via a mechanism that modulates CD14 trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO18A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MYO18A
Gene Accession No. Q92614, Q9JMH9
Gene Alias CD245; KIAA0216; MAJN; molecule associated with JAK3 N-terminus; Myo18a; MyoPDZ; myosin 18a; myosin containing a PDZ domain; myosin containing PDZ domain; myosin XVIIIa; myosin XVIIIb; myosin-XVIIIa; MysPDZ; SP-A receptor subunit SP-R210 alphaS; SPR210; SP-R210; Surfactant protein receptor SP-R210; Unconventional myosin-XVIIIa
Gene Symbols MYO18A
Host Species Rabbit
Immunogen A synthesized peptide derived from human MYO18A(Accession Q92614), corresponding to amino acid residues A958-T1008.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360013, 360570, 399687
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.