missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mycobacterium tuberculosis GlcB Polyclonal Antibody
GREENER_CHOICE

Product Code. 17703724 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17703724 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17703724 Supplier Invitrogen™ Supplier No. PA5122322

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody reacts with Mycobacterium tuberculosis protein GlcB (malate synthase, Rv1837c).

Involved in the glycolate utilization. Catalyzes the condensation and subsequent hydrolysis of acetyl-coenzyme A (acetyl-CoA) and glyoxylate to form malate and CoA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycobacterium tuberculosis GlcB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene glcB
Gene Accession No. P9WK17
Gene Symbols glcB
Host Species Rabbit
Immunogen Recombinant GlcB protein (produced in E. coli).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 885713
Target Species Bacteria
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.