Learn More
Lonza™ Mycoalert™ Mycoplasma Detection Kit
Product Code. p-8010791
Click to view available options
Quantity:
100 Tests
25 Tests
50 Tests
Unit Size:
1 set
25 tests
50 tests
Product Code. 11650261
Brand: Lonza™ LT07218
Detect mycoplasma in culture. Mycoalert™ Mycoplasma Detection Kit is a selective biochemical test that exploits the activity of mycoplasmal enzymes which are found in all six of the main mycoplasma cell culture contaminants and the vast majority of 180 mycoplasma species, but are not present in eukaryotic cells.
Convenient & fast
  • Simply add two reagents to the culture supernatant
  • Perform two luminescence readings
  • Results in < 20 min
  • Easy interpretation of results
Universal
  • Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma)
  • Negligible interference with media components
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Mycoalert reagent: 5 x 600μL, Mycoalert assay buffer: 1 x 10mL, Mycoalert substrate: 5 x 600μL
2 to 8°C
Cell Type Mammalian Cells
Product Type Mycoplasma Detection
Quantity 25 Tests
compliance-icons compliance-icons
Signal Word
  • Danger||
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Acetyl dihydrogen phosphate, lithium potassium salt, Dilithium carbamoyl phosphate hydrate, EDTA Dis