Learn More
Cytiva (formerly Pall Lab) Mustang Q XT Acrodisc Unit
Click to view available options
:
0.8 μm
Description
- Fast, easy-to-use, scale-down Mustang membrane unit facilitates process development work
- Device volume of <1 mL minimizes sample volume requirements
- 16 layers of Mustang membrane allows easy scale-up
Specifications
Specifications
| Bed Volume | 0.86 mL |
| Material (Membrane) | PES |
| Pore Size | 0.8 μm |
| Sterility | Non-sterile |
| Inlet Style | Female Luer Lock |
| Outlet Style | Female Luer Lock |