missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscone, ≥95%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
Specifications
Specifications
| Color | Colorless |
| Density | 0.84g/mL |
| Assay Percent Range | ≥95% |
| Quantity | 250 mg |
| Formula Weight | 238.43 |
| Physical Form | Liquid |
| Chemical Name or Material | Muscone |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction