Learn More
Benchmark Scientific MultiTherm™ Shaker with Heating and Cooling
Product Code. 16440251
Change view
Click to view available options
Speed Range:
200 to 1500 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range unitSize
16440251 200 to 1500 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16440251 Supplier Benchmark Scientific Supplier No. H5000HCE
Only null left
Add to basket
Add to basket
The MultiTherm™ is a temperature controlled, variable speed vortexer that heats and cools. Suitable for a wide range of applications, the MultiTherm™ accepts tubes from 0.2 to 50 mL in exchangeable blocks.
  • Temperature control from 0° to 100°C
  • Exchangeable blocks (sold separately) for tubes from 0.20 - 50 mL
  • Precision contoured wells for uniform thermal transfer
  • Sequence link function for linking or repeating programs
  • Temperature and shaking functions are independent of each other
Specifications
Orbit 3 mm
Speed Range 200 to 1500 rpm
Voltage 240 V
Temperature Range (Metric) -20°C to +100°C
Wattage 500 W
Amperage 1.5 A
Temperature Range (English) −68°F to +212°F
Frequency 50 Hz
Material ABS
Height (English) 8.25 in.
Height (Metric) 21 cm
Width (English) 8.25 in.
Width (Metric) 21 cm
Shipping Weight (English) 22 lb.
Shipping Weight (Metric) 10 kg
Model MultiTherm
Humidity Range >80%
Timer Range 99:59 hr.