missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Multiparameter Chemical Test Kit

Product Code. 11666790
Click to view available options
For Use With (Application):
Cooling and Boiler Monitoring
Environmental Monitoring
Marine/Aquaculture Monitoring
NPK Soil Testing
Water Quality Monitoring
Unit Size:
Each
Pack of 100
This item is not returnable. View return policy

Product Code. 11666790

Brand: Hanna Instruments™ HI3896

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Multiparameter Chemical Test Kit features pre-made, pre-measured titration reagents for the determination of multiple parameters commonly measured for environmental/facility monitoring.

These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.

  • Complete setup: All required materials are included, such as beakers, syringes, a case and pre-measured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted

Specifications

For Use With (Application) NPK Soil Testing
For Use With (Equipment) Soil samples
Includes All necessary reagents and accessories for each test, such as the test tubes, spoon, brush, color comparison cards, reagent packets and solution bottles, and a handbook and hard carrying case
No. of Tests 4 parameters x 25 tests
Quantity 1 Kit
Range Various
Sufficient For Testing of: pH, nitrogen (N), phosphorus (P), and potassium (K)
Test Method Colorimetry and turbidimetry
Type Chemical Test Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.