Learn More
Gilson™ PIPETMAN™ M Multi-Channel
GREENER_CHOICE
Product Code. 16378157 Shop All Gilson Products
Change view
Click to view available options
Number of Channels:
8
12
Volume (Metric):
0.5 to 10 μL
1 to 20 μL
10 to 100 μL
10 to 300 μL
20 to 200 μL
50 to 1200 μL
Unit Size:
Each
Product Code. Number of Channels Volume (Metric) unitSize
16378157 12 10 to 100 μL Each
16328157 8 0.5 to 10 μL Each
16338157 12 0.5 to 10 μL Each
16348157 8 1 to 20 μL Each
16358157 12 1 to 20 μL Each
16368157 8 10 to 100 μL Each
16388157 8 20 to 200 μL Each
16398157 12 20 to 200 μL Each
16308167 8 10 to 300 μL Each
16318167 12 10 to 300 μL Each
16328167 8 50 to 1200 μL Each
16338167 12 50 to 1200 μL Each
12 options
Product Code. 16378157
Brand: Gilson™ F81053
PIPETMAN™ M Multi-Channel is a Bluetooth™-enabled electronic pipette that offers accuracy and precision in both standard and repetitive pipetting modes.
  • Allows accurate and precise pipetting
  • The motorized piston allows for high pipetting reproducibility and accuracy in all modes
  • Associated with TRACKMAN™ Connected and its PipettePilot™ application, PIPETMAN™ M Connected reduces the risk of human error by transferring verifiable pipetting parameters and commands in the pipette
  • Increased functionality
  • The intuitive interface has five pipetting modes for a large number of applications: pipette, repetitive, mix, reverse, and custom
  • Create and transfer up to 10 custom protocols from PIPETMAN™ M Connected Software into your pipette
  • Bluetooth™ connection enables communication with TRACKMAN™ Connected and helps guide you through your protocol telling you where, when, and how much to pipette with real-time interactions
  • Simple and comfortable to use
  • Aspirate and dispense with one click of the push button and eject tips with ease
  • The lightweight and ergonomic design rests comfortably in your hand for maximum pipetting comfort
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Universal
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Orange
Disposable Reusable
For Use With Pipette Tip, TRACKMAN™ Connected
Grip Type Ergonomic
Material Polypropylene
Model P12X100M BT
Number of Channels 12
Product Line Pipetman M
Programmability Yes
Sterility Non-sterile
Type Pipette
Volume (Metric) 10 to 100 μL
For Use With (Equipment) Pipette Tips, TRACKMAN™ Connected
Quantity 1
Warranty 1 Year
Length (English) 15.75 in.
Length (Metric) 32.4 cm
Width (English) 5.9 in.
Width (Metric) 15 cm
Height (English) 2.16 in.
Height (Metric) 5.5 cm
Electrical Requirements 3.7 V and 420 mAh (1.55 Wh)
Fit for Purpose Certified Quality