missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mtRNA polymerase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35578-20ul
This item is not returnable.
View return policy
Description
mtRNA polymerase Polyclonal antibody specifically detects mtRNA polymerase in Human,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| mtRNA polymerase | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| APOLMTEC 2.7.7.6, DNA-directed RNA polymerase, mitochondrial, h-mtRPOL, MTRNAP, MTRPOL, polymerase (RNA) mitochondrial (DNA directed) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 42-230 of human mtRNA polymerase (NP_005026.3).,, Sequence:, SSASPQEQDQDRRKDWGHVELLEVLQARVRQLQAESVSEVVVNRVDVARLPECGSGDGSLQPPRKVQMGAKDATPVPCGRWAKILEKDKRTQQMRMQRLKAKLQMPFQSGEFKALTRRLQVEPRLLSKQMAGCLEDCTRQAPESPWEEQLARLLQEAPGKLSLDVEQAPSGQHSQAQLSGQQQRLLAFF | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5442 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction