missing translation for 'onlineSavingsMsg'
Learn More

MTIF3 Monoclonal Antibody (OTI4G2), TrueMAB™, OriGene

Product Code. 17578222
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17578222 30 μL 30µL
17558222 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17578222 Supplier Origene Technologies Supplier No. TA800420S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MTIF3 Monoclonal antibody specifically detects MTIF3 in Canine, Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MTIF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene EPHX1
Gene Accession No. P07099, Q9D379
Gene Alias 2810012L14Rik; AI414549; DC38; IF3(mt); IF-3(Mt); IF3mt; IF-3Mt; mitochondrial translational initiation factor 3; Mtif3; Translation initiation factor IF-3, mitochondrial
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 21-230 of human EPHX1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13849, 2052, 480113
Target Species Canine, Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.