missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Msx3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17127668 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17127668 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17127668 Supplier Invitrogen™ Supplier No. PA5116430

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Msx3 Polyclonal Antibody for Western Blot

Involved in the development of the inner ear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Msx3
Applications Western Blot
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Msx3
Gene Accession No. Q01703
Gene Alias AI323377; etID309953.13; homeo box, msh-like 3; homeobox; homeobox protein MSH-C; homeobox protein MSX-3; homeobox, msh-like 3; msh homeobox 3; mshC; msh-C; Msx3; Msx-3; msxc; msx-c; muscle segment homeobox 3; muscle segment homeobox C; zgc:86596
Gene Symbols Msx3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 123-157 amino acids from the Central region of DANRE msxc.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30526
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.