missing translation for 'onlineSavingsMsg'
Learn More

MS023, MedChemExpress

Product Code. p-200097198
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487646

Brand: Medchem Express HY19615

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MS023 is a potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs) inhibitor, with IC50s of 30, 119, 83, 4 and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6, and PRMT8, respectively.

MS023 is a potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs) inhibitor, with IC50s of 30, 119, 83, 4 and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6, and PRMT8, respectively.

Specifications

Chemical Name or Material MS023
CAS 1831110-54-3
Color Light Nude
Purity Grade Notes Research
Molecular Formula C17H25N3O
Quantity 100 mg
Solubility Information H2O : 66.67 mg/mL (231.98 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (104.38 mM)
SMILES CN(CCN)CC1=CNC=C1C2=CC=C(OC(C)C)C=C2
Molecular Weight (g/mol) 287.4
Formula Weight 287.4
Percent Purity 99.12%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.