Learn More
Ismatec™ MS-CA 2/660 Base-plate Cassette Pump
Click to view available options
Flow Rate:
0.083 to 26 mL/min.
Unit Size:
Each
Specifications
Specifications
| Certifications/Compliance | CE Compatibility (EN 61326-1/EN 61010-1) |
| Description | 3 stop color coded |
| Control Type | Fixed-Speed |
| Display Type | No Display |
| Enclosure Rating | IP30 |
| Flow Rate | 0.083 to 26 mL/min. |
| Maximum Pressure | 14.5 psi (Differential) |
| Motor Speed | 60 rpm |
| Reversible | Yes |
| Includes | Click-n-go Cassettes, Operating Manual, Power Cord |
| Show More |