missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRT68921 dihydrochloride, MedChemExpress
Description
MRT68921 dihydrochloride is a potent inhibitor of ULK1 and ULK2, with IC50 values of 2.9 nM and 1.1 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | MRT68921 dihydrochloride |
| CAS | 2080306-21-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H36Cl2N6O |
| Quantity | 50 mg |
| Solubility Information | H2O : 31.25 mg/mL (61.58 mM; Need ultrasonic) ∣DMSO : 8.33 mg/mL (16.41 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | [H]Cl.O=C(C1CCC1)NCCCNC2=NC(NC3=CC4=C(CN(C)CC4)C=C3)=NC=C2C5CC5.[H]Cl |
| Molecular Weight (g/mol) | 507.5 |
| Formula Weight | 507.5 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?