Learn More
Tocris Bioscience™ MRT 67307 dihydrochloride
GREENER_CHOICE
Salt inducible kinase (SIK) inhibitor
Brand:  Tocris Bioscience™ 5134/10
Encompass
Product Code. 15786625
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
MRT 67307 dihydrochloride is a salt inducible kinase (SIK) inhibitor (IC50 values are 67, 250 and 430 nM for SIK2, SIK1 and SIK3 respectively). Also inhibits TBK1, MARK1-4, IKKε and NUAK1 (IC50 values are 19, 27-52, 160 and 230 nM respectively). Has no effect on IKKα or IKKβ. Induces IL-10 secretion and inhibits TNF-α and IL-6 secretion in bacterial LPS-stimulated macrophages. Also enhances IL-1-induced activation of NFκB-dependent gene transcription in mouse embryonic fibroblast (MEF) cells. Inhibits autophagy.
Specifications
Specifications
>98%
10 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions