Learn More
Tocris Bioscience™ MRS 2905
Potent and selective P2Y14 receptor agonist
Brand: Tocris Bioscience™ 5633/1
This item is not returnable.
View return policy
Description
MRS 2905 is a potent and selective P2Y14 receptor agonist (EC50 = 0.92 nM). Exhibits >2000-fold selectivity over P2Y6.Specifications
| 2-Thiouridine-5′-O-(α, β-methylene)diphosphate trisodium salt | |
| 1 mg |