Learn More
Tocris Bioscience™ MRS 2905
GREENER_CHOICE
Potent and selective P2Y14 receptor agonist
Brand:  Tocris Bioscience™ 5633/1
Encompass
Product Code. 15786745
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
MRS 2905 is a potent and selective P2Y14 receptor agonist (EC50 = 0.92 nM). Exhibits >2000-fold selectivity over P2Y6.
Specifications
Specifications
2-Thiouridine-5′-O-(α, β-methylene)diphosphate trisodium salt
1 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions