missing translation for 'onlineSavingsMsg'
Learn More

MRS 2578, MedChemExpress

Product Code. p-200098944
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16491707

Brand: Medchem Express HY13104

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MRS 2578 is a selective and potent P2Y6 receptor antagonist with IC50s of 37 nM (human) and 98 nM (rat). MRS 2578 exhibits insignificant activity at P2Y1, P2Y2, P2Y4, and P2Y11 receptors.

MRS 2578 is a selective and potent P2Y6 receptor antagonist with IC50s of 37 nM (human) and 98 nM (rat). MRS 2578 exhibits insignificant activity at P2Y1, P2Y2, P2Y4, and P2Y11 receptors.

Specifications

Chemical Name or Material MRS 2578
Physical Form Solid
CAS 711019-86-2
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H20N6S4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (105.78 mM; Need ultrasonic)
SMILES S=C(NCCCCNC(NC1=CC(N=C=S)=CC=C1)=S)NC2=CC(N=C=S)=CC=C2
Molecular Weight (g/mol) 472.67
Formula Weight 472.67
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.