Learn More
MRS 2578, MedChemExpress
Product Code. 16491707
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16491707
Brand: Medchem Express HY13104
MedChemExpress MRS 2578 is a selective and potent P2Y6 receptor antagonist with IC50s of 37 nM (human) and 98 nM (rat). MRS 2578 exhibits insignificant activity at P2Y1, P2Y2, P2Y4, and P2Y11 receptors.
MRS 2578 is a selective and potent P2Y6 receptor antagonist with IC50s of 37 nM (human) and 98 nM (rat). MRS 2578 exhibits insignificant activity at P2Y1, P2Y2, P2Y4, and P2Y11 receptors.
Specifications
Chemical Name or Material MRS 2578
Physical Form Solid
CAS 711019-86-2
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H20N6S4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (105.78 mM; Need ultrasonic)
SMILES S=C(NCCCCNC(NC1=CC(N=C=S)=CC=C1)=S)NC2=CC(N=C=S)=CC=C2
Molecular Weight (g/mol) 472.67
Formula Weight 472.67
Percent Purity 98.0%
Grade Research