missing translation for 'onlineSavingsMsg'
Learn More

MRPS27 Monoclonal Antibody (OTI3B9), TrueMAB™, OriGene

Product Code. 17597812
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17597812 30 μL 30µL
17577812 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17597812 Supplier Origene Technologies Supplier No. TA800199S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRPS27 Monoclonal antibody specifically detects MRPS27 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MRPS27
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Mrps27
Gene Accession No. Q92552
Gene Alias 2610028H14Rik; 28S ribosomal protein S27, mitochondrial; KIAA0264; mitochondrial 28S ribosomal protein S27; mitochondrial ribosomal protein S27; Mitochondrial small ribosomal subunit protein mS27; mrps27; MRP-S27; S27mt
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 151-414 of human MRPS27 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23107
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.