Learn More
Invitrogen™ MRPL46 Polyclonal Antibody

Description
Immunogen sequence: MAAPVRRTLL GVAGGWRRFE RLWAGSLSSR SLALAAAPSS NGSPWRLLGA LCLQRPPVVS KPLTPLQEEM ASLLQQIEIE RSLYSDHELR ALDENQRLAK KKADLHDEED EQDILLAQDL EDMWEQKFLQ FKLGARITEA DEKNDRTSLN RKLDRNLVLL VREKFGDQDV WILPQAEWQP GETLRGTAER TLATLSENNM EAKFLGNAPC GHYTFKFPQA MRTESNLGAK VFFFKALLLT GDFSQAGNKG HHVWVTKDEL GDYLKPKYLA QVRRFVSDL; Positive Samples: Mouse brain; Cellular Location: Mitochondrion.
Specifications
Specifications
| Antigen | MRPL46 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.57 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | MRPL46 |
| Gene Accession No. | Q9EQI8, Q9H2W6 |
| Gene Alias | 3110052F15Rik; 39S ribosomal protein L46, mitochondrial; C15orf4; C80107; L46mt; LIECG2; Mitochondrial large ribosomal subunit protein mL46; mitochondrial ribosomal protein L46; Mrpl46; MRP-L46; P2ECSL |
| Gene Symbols | MRPL46 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.