missing translation for 'onlineSavingsMsg'
Learn More

MRPL15 Monoclonal Antibody (OTI7A3), TrueMAB™, OriGene

Product Code. 17507803
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17507803 30 μL 30µL
17587793 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17507803 Supplier Origene Technologies Supplier No. TA807342S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRPL15 Monoclonal antibody specifically detects MRPL15 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MRPL15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PDX1
Gene Accession No. P52945
Gene Alias 39S ribosomal protein L15, mitochondrial; HSPC145; L15mt; Mitochondrial large ribosomal subunit protein uL15m; mitochondrial ribosomal protein L15; Mrpl15; MRP-L15; MRP-L7; ribosomal protein, mitochondrial, L7; RPML7
Host Species Mouse
Immunogen Full length human recombinant protein of human PDX1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3651
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.