missing translation for 'onlineSavingsMsg'
Learn More

MRK-016, MedChemExpress

Product Code. 16499536
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499536

Brand: Medchem Express HY100370

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MRK-016 is a selective, orally bioavailable inverse agonist of GABAA α5 receptor, with an EC50 of 3 nM for GABAA α5, and Kis of 0.83, 0.85, 0.77 and 1.4 nM for human GABAA α1β3γ2, GABAA α2β3γ2, GABAA α3β3γ2, and GABAA α5β3γ2, respectively; MRK-016 also readily penetrates the CNS.

MRK-016 is a selective, orally bioavailable inverse agonist of GABAA α5 receptor, with an EC50 of 3 nM for GABAA α5, and Kis of 0.83, 0.85, 0.77 and 1.4 nM for human GABAA α1β3γ2, GABAA α2β3γ2, GABAA α3β3γ2, and GABAA α5β3γ2, respectively

Specifications

Chemical Name or Material MRK-016
Physical Form Powder
CAS 342652-67-9
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H20N8O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (135.73 mM; Need ultrasonic)
SMILES CN1N=CN=C1COC2=NN3C(C4=NOC(C)=C4)=NN=CC3=C2C(C)(C)C
Molecular Weight (g/mol) 368.39
Formula Weight 368.39
Percent Purity 98.29%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.