missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRGPRG Polyclonal Antibody
GREENER_CHOICE

Product Code. 16688574
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16688574 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16688574 Supplier Invitrogen™ Supplier No. PA5102104

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MRGRG.

Orphan receptor. May regulate nociceptor function and/or development, including the sensation or modulation of pain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRGPRG
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Mrgprg
Gene Accession No. Q86SM5
Gene Alias G protein-coupled receptor 169; G protein-coupled receptor MRGG; GPR169; G-protein coupled receptor 169; MAS related GPR family member G; MAS-related gene G; MAS-related GPR, member G; mas-related G-protein coupled receptor member G; MAS-related G-protein coupled receptor, member G; Mrgg; MRGPRG
Gene Symbols Mrgprg
Host Species Rabbit
Immunogen A synthesized peptide derived from human MRGPRG(Accession Q86SM5), corresponding to amino acid residues F231-G281.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 386746
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.