missing translation for 'onlineSavingsMsg'
Learn More

Mrc2 Antibody (Quark patent anti-ENDO 180) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30172610
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30172610 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30172610 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328883JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

Mrc2 Monoclonal antibody specifically detects Mrc2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mrc2
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone Quark patent anti-ENDO 180
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias CD280 antigen, CD280C-type mannose receptor 2, CLEC13EUPAR-associated protein, ENDO180C-type lectin domain family 13 member E, Endocytic receptor 180, KIAA0709FLJ35911, Macrophage mannose receptor 2, mannose receptor, C type 2, UPARAPendocytic receptor (macrophage mannose receptor family), urokinase plasminogen activator receptor-associated protein, Urokinase receptor-associated protein, Urokinase-type plasminogen activator receptor-associated protein
Host Species Human
Immunogen MRC2/ CD280
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 9902
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.