missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPV17L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560245
This item is not returnable.
View return policy
Description
Immunogen sequence: GCLEGQTVGE SCQELREKFW EFYKADWCVW PAAQFVNFLF VPPQFRVTYI NGLTLGWDTY LSYLKYRSPV PLTPPGCVAL DTRAD Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
FKSG24 is a multi-pass membrane proteinPotential. It belongs to the peroxisomal membrane protein PXMP2/4 family. The exact function of FKSG24 remains unknown.
Specifications
| MPV17L2 | |
| Polyclonal | |
| Unconjugated | |
| MPV17L2 | |
| FKSG24; MPV17 mitochondrial inner membrane protein like 2; MPV17 mitochondrial membrane protein-like 2; MPV17L2; mpv17-like protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84769 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q567V2 | |
| MPV17L2 | |
| Recombinant protein corresponding to Human MPV17L2. Recombinant protein control fragment (Product #RP-99025). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction