missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPND Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24420-1-AP-150UL
This item is not returnable.
View return policy
Description
Probable protease.Specifications
| MPND | |
| Polyclonal | |
| Unconjugated | |
| MPND | |
| MPN domain containing, MPN domain containing protein, MPND | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 68047, 84954 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q3TV65, Q8N594 | |
| MPND | |
| MPND Fusion Protein Ag19531 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction