missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPND Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
484.00 EUR
Specifications
| Antigen | MPND |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
MPND Polyclonal specifically detects MPND in Human samples. It is validated for Western Blot.Specifications
| MPND | |
| Polyclonal | |
| Purified | |
| RUO | |
| EC 3.4, FLJ14981, MPN domain containing, MPN domain-containing protein | |
| MPND | |
| IgG | |
| Protein A purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| NP_116257 | |
| 84954 | |
| Synthetic peptide directed towards the N terminal of human MPNDThe immunogen for this antibody is MPND. Peptide sequence MAAPEPLSPAGGAGEEAPEEDEDEAEAEDPERPNAGAGGGRSGGGGSSVS. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title