Learn More
Welch Ilmvac™ MPC 301 Z Vacuum Pump
Shop All Welch Ilmvac Products
Click to view available options
Flow Rate:
41 L/min.
Unit Size:
Each
Description
Use these pumps for chemical duty applications. Welch Ilmvac™ MPC 301 Z Vacuum Pumps use PTFE and other fluorinated plastics for the wetted parts to allow aggressive solvent and acid vapors to be pumped.
- Low weight and excellent ergonomics
- Proven diaphragm technology offers a double benefit to the user
- ON/OFF switch and internal protective thermal switch for the motor, mains cable and plug
- Carry handle
- Vibration isolating feet
- MP models come with and exhaust silencer
- MPC models come with a gas ballast valve
- analytically pure, oil free vacuum
- User friendly
- Light weight, rugged design
- Designed for permanent operation
- Maintenance-free drive system and proven long diaphragm life
- Minimal operation vibration
- Wide vacuum and flow range to match application
- Superior chemical resistance (MPC models)
Specifications
Specifications
| Max. Vacuum | 29.7 in.Hg |
| Dimensions (L x W x H) | 23 x 26.5 x 16.9 cm |
| Wattage | 180 w |
| Weight (Metric) | 11.2 kg |
| Flow Rate | 41 L/min. |
| Pump Type | Vacuum Pump |
| Enclosure Rating | IP54 |
| Applications | Chemical Duty |
| Maximum Pressure | 8 mbar |