missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ SD Medium-LEU, Powder
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 x 500 mL
10 x 1000 mL
Unit Size:
10 L
5 L
Description
- Each SD formulation contains the appropriate CSM-based amino acid mixture and 1.7g/liter YNB, 5g/liter Ammonium Sulfate, and 20g/liter Dextrose
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Specifications
| Certifications/Compliance | Molecular biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Product Type | Yeast Media |
| Formulation | YNB 1.7g/L, Ammonium Sulfate 5g/L, Dextrose 20g/L, CSM-Leu 0.69g/L |
| For Use With (Application) | For confirming auxotrophic phenotypes and selecting Yeast transformants |
| Includes | All necessary components for selective growth of S. cereviseae |
| Packaging Type | Pouch |
| Quantity | 10 x 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction