missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Protexidase Peroxidase Protective Buffer

Product Code. 11434346 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434346 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434346 Supplier MP Biomedicals™ Supplier No. 08980631
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Stabilizes high dilutions of HRP-antibody conjugates

  • Peroxidase Protective Buffer which stabilizes HRP conjugates.
  • This protective buffer is compatible with common blocking agents such as BSA, skim milk (Blotto), Tween-20 and normal serum.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 mL
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.