Learn More
MP Biomedicals™ Poly-L-Glutamic Acid, MW 50,000-100,000
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mg
1000 mg
Unit Size:
1g
500mg
Specifications
Specifications
| Purity | 95% |
| Content And Storage | 0°C |
| Format | Powder |
| For Use With (Application) | Used in the construction of Polypeptide based Copolymer membranes, micelles and nanoparticles |
| Molecular Weight (g/mol) | 50000 to 100000 |
| Product Type | Poly-L-Glutamic Acid from Synthetic |
| Quantity | 500 mg |