Learn More
MP Biomedicals™ MT Buffer
Product Code. 12877860 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
8 mL
Unit Size:
8mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12877860 8 mL 8mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12877860 Supplier MP Biomedicals™ Supplier No. 116511202
MT Buffer is a lysis solution, carefully developed to protect and solubilize nucleic acids and proteins upon cell lysis.
  • MT Buffer is a lysis solution component included in the FastRNAPro Soil Direct and Indirect Kits, the FastDNA Spin Kit for Feces, and the FastDNA 50 mL Spin Kit for Soil.
  • Used as a stabilization agent during sample lysis.
  • Homogenization takes place in the presence of MT Buffer, this reagent is carefully developed to protect and solubilize nucleic acids and proteins upon cell lysis.
  • Thus allowing extraction of genomic DNA with minimal RNA contamination.
TRUSTED_SUSTAINABILITY
Specifications
Sample Type Feces, Soil, Wastewater, Environmental Water, Sludge
For Use With (Equipment) FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1
For Use With (Application) Sample preparation, sample lysis, nucleic acid isolation and purification, molecular biology, gene expression, microbiological analysis, gene function, gene regulation
Product Type MT Buffer
Quantity 8 mL
Sample Volume Up to 500 mg Per Prep
Sample Source Microorganisms
compliance-icons
Product Identifier
  • MT Buffer
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium dodecyl sulphate
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.