missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Chemical Name or Material | Earle's Balanced Salt |
| Color | Clear |
| Concentration | 1 X |
| pH | 7.26 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Formulation | Calcium Chloride Dihydrate 264.90mg/L, Magnesium Sulfate 97.7mg/L, Potassium Chloride 400mg/L, Sodium Bicarbonate 2200mg/L, Sodium Chloride 6800mg/L, Sodium Phosphate Monobasic Monohydrate 140mg/L, Dextrose 1000mg/L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?