Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Free Fatty Acid, Stem Cell Culture
Bovine serum albumin suitable for stem cell culture and media formulation requiring very low levels of lipids
Brand: MP Biomedicals™ 0219989925
Additional Details : CAS Number : 9048-46-8 Weight : 0.02500kg
Description
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Features low endotoxin, low level of lipids, fatty acids and hormones, e.g. insulin, T3, T4.
Physical characteristics:
- Form: Off white to brown powder
- Purity: ≥98% (w/w)
- Total Protein: ≥96%
- Free Fatty Acids ≤0.005% w/w
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Insulin, T3, T4
Specifications
Mammalian Cell Culture | |
CellMaxx™ Bovine Albumin | |
Chromatography | |
Bovine | |
Powder |
66 | |
6.5 to 7.5 | |
25g | |
4°C |