missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Free Fatty Acid, Stem Cell Culture
Bovine serum albumin suitable for stem cell culture and media formulation requiring very low levels of lipids
79.55 EUR - 3142.90 EUR
Specifications
For Use With (Application) | Mammalian Cell Culture |
---|---|
Molecular Weight (g/mol) | 66 |
Name | CellMaxx™ Bovine Albumin |
pH Range | 6.5 to 7.5 |
Purification Method | Chromatography |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17338573
|
MP Biomedicals™
0219989910 |
10g |
79.55 EUR
10g |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17378583
|
MP Biomedicals™
0219989925 |
25g |
129.32 EUR
25g |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11401665
|
MP Biomedicals™
0219989980 |
100g |
375.24 EUR
100g |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11411665
|
MP Biomedicals™
0219989990 |
500g |
1680.10 EUR
500g |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11421665
|
MP Biomedicals™
0219989991 |
1000g |
3142.90 EUR
1kg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Features low endotoxin, low level of lipids, fatty acids and hormones, e.g. insulin, T3, T4.
Physical characteristics:
- Form: Off white to brown powder
- Purity: ≥98% (w/w)
- Total Protein: ≥96%
- Free Fatty Acids ≤0.005% w/w
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Insulin, T3, T4
Specifications
Mammalian Cell Culture | |
CellMaxx™ Bovine Albumin | |
Chromatography | |
4°C |
66 | |
6.5 to 7.5 | |
Bovine | |
Powder |