missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ (Arg8)-Vasopressin

Product Code. 11444091 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 11444091

Brand: MP Biomedicals™ 0215296201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • Contains a disulfide bond between Cys1 and Cys6

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Biology and Analysis
Molecular Weight (g/mol) 1084.23
Name (Arg-8)-Vasopressin (AVP)
Quantity 1 mg
Storage Requirements -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.