missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Aprotinin Protein

Product Code. 11410662 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410662 5 mg 5mg
11420662 25 mg 25mg
11430662 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410662 Supplier MP Biomedicals™ Supplier No. 0219410105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From bovine lung

Protease Inhibitor. Activity: ≥3 TIU/mg. Unit Definition: One trypsin inhibitor unit (TIU), which is equal to 900 kallikrein inhibitor units (KIU) and 8800 Schwert and Takenake units (STU), is the amount of inhibitor per mg which hydrolyzes one μmol of benzoyl-DL-arginine-p-nitroanilide (BAPNA) per minute. One TIU reduces the activity of two trypsin units by 50%. It is reactive against trypsin, chymotrypsin, plasmatic and glandular kininogenases, plasmin, urokinase, clotting factor XIIa, protein C, proteinases of the complement system, and leukocyte and tissue proteinases.

TRUSTED_SUSTAINABILITY

Specifikationer

For Use With (Application) Molecular Biology
Molecular Weight (g/mol) 6511.23
Name Aprotinin from Bovine Lung
pH Range 5 to 7
Quantity 5 mg
Source Bovine Lung
Storage Requirements 4°C
Form Lyophilized
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.